missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Letrozole, MedChemExpress
Description
Letrozole (CGS 20267) is a potent, selective, reversible and orally active non-steroidal inhibitor of aromatase, with an IC50 of 11.5 nM. Letrozole selective inhibits estrogen biosynthesis, and can be used for the research of breast cancer.
Specifications
Specifications
| Chemical Name or Material | Letrozole |
| Physical Form | Powder |
| CAS | 112809-51-5 |
| Color | Off-White |
| Quantity | 200 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C17H11N5 |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | CGS 20267 |
| Solubility Information | DMSO : 50 mg/mL (175.25 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?