missing translation for 'onlineSavingsMsg'
Learn More

Letrozole, Dr. Ehrenstorfer

Product Code. 30325420
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30325420 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325420 Supplier Dr. Ehrenstorfer Supplier No. DREC14628500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Letrozole
CAS 112809-51-5
Analyte or Component Names Letrozole
Shipping Condition +20°C
Molecular Formula C17 H11 N5
Certifications/Compliance ISO/IEC 17025
Synonym Letrozole
InChI Formula InChI=1S/C17H11N5/c18-9-13-1-5-15(6-2-13)17(22-12-20-11-21-22)16-7-3-14(10-19)4-8-16/h1-8,11-12,17H
SMILES N#Cc1ccc(cc1)C(c2ccc(cc2)C#N)n3cncn3
IUPAC Name 4-[(4-cyanophenyl)-(1,2,4-triazol-1-yl)methyl]benzonitrile
Molecular Weight (g/mol) 285.3
Quantity 50 mg
Formula Weight 285.1014 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.