missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LETMD1 Polyclonal Antibody

Description
Immunogen sequence: FPRQLLIRHFW TPKQQTDFLD IYHAFRKQSH PEIISYLEKV IPLISDAGLR WRLTDLCTKI QRG Highest antigen sequence identity to the following orthologs - mouse 59%, rat 66%.
HCCR-1 (human cervical cancer oncogene 1), also known as HCCR-2 or LETMD1, is a 360 amino acid single-pass membrane protein that is expressed in the kidney, liver, skeletal muscle, heart and brain. It is suggested that HCCR-1 may be involved in tumorigenesis and may function as a negative regulator of the p53 tumor suppressor. Overexpression of HCCR-1 may cause mitochondrial dysfunction that can lead to UVC or staurosporine-induced apoptosis resistance and progression of tumor formation. HCCR-1 is considered a candidate biomarker for breast cancer. Various human tumors including leukemia, lymphoma, and carcinomas of the breast, kidney, ovary, stomach, colon, and uterine cervix have high HCCR-1. Six isoforms exist due to alternative splicing events.
Specifications
Specifications
| Antigen | LETMD1 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | LETMD1 |
| Gene Accession No. | Q6P1Q0 |
| Gene Alias | 1110019O13Rik; AI593524; BB130465; BB235638; Cervical cancer 1 proto-oncogene protein p40; cervical cancer proto-oncogene 2 protein; Cervical cancer receptor; HCCR1; HCCR-1; HCCR-2; HCRR-2; LETM1 domain containing 1; LETM1 domain-containing protein 1; LETMD1; MCC-32; Mccr; regulator of TP53; RGD1310591 |
| Gene Symbols | LETMD1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?