missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LETM2 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97437MFV610
This item is not returnable.
View return policy
Description
LETM2 Polyclonal antibody specifically detects LETM2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| LETM2 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| FLJ25409, LETM1 and EF-hand domain-containing protein 2, LETM1 domain-containing protein LETM2, mitochondrial, leucine zipper-EF-hand containing transmembrane protein 1-like protein, leucine zipper-EF-hand containing transmembrane protein 2, Leucine zipper-EF-hand-containing transmembrane protein 1-like | |
| Produced in rabbits immunized with E. coli-derived Human LETM2 fragment. | |
| 0.1 mL | |
| Endocrinology | |
| 137994 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction