missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lestaurtinib, MedChemExpress
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
Lestaurtinib (CEP-701) is an orally active and selective RPTKs (receptor protein tyrosine kinase) inhibitor, competitively inhibits ATP binding to the TrkA/B/C domain. Lestaurtinib inhibits RPTKs phosphorylation, with IC50s of 2, 25 and 0.9 nM for FLT3, TrkA and JAK2, respectively. Lestaurtinib induces apoptosis and cycle arrest, also can inhibit growth of tumor.
Specifications
Specifications
| Chemical Name or Material | Lestaurtinib |
| CAS | 111358-88-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C26H21N3O4 |
| Quantity | 5 mg |
| Synonym | CEP-701 KT-5555 |
| Solubility Information | DMSO : 50 mg/mL (113.78 mM; Need ultrasonic) |
| SMILES | O=C(NC1)C2=C1C3=C(C4=C52)N(C6=C3C=CC=C6)[C@@]([C@@](O)(CO)C7)(C)O[C@H]7N4C8=C5C=CC=C8 |
| Molecular Weight (g/mol) | 439.46 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction