missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lesinurad sodium, MedChemExpress
Description
Lesinurad sodium is a URAT1 and OAT inhibitor, is determined to be a substrate for the kidney transporters OAT1 and OAT3 with Km values of 0.85 and 2 μM, respectively.
Specifications
Specifications
| Chemical Name or Material | Lesinurad sodium |
| Physical Form | Solid |
| CAS | 1151516-14-1 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C17H13BrN3NaO2S |
| For Use With (Application) | Metabolism-protein/nucleotide metabolism |
| Synonym | RDEA-594 sodium |
| Solubility Information | DMSO : ≥ 26 mg/mL (61.00 mM) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?