missing translation for 'onlineSavingsMsg'
Learn More

Lercanidipine hydrochloride, MedChemExpress

Product Code. 16401436
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16401436 200 mg 200mg
16488386 100 mg 100mg
16474417 50 mg 50mg
16428558 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16401436 Supplier Medchem Express Supplier No. HYB0612A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lercanidipine hydrochloride is a lipophilic third-generation dihydropyridine-calcium channel blocker (DHP-CCB). Lercanidipine hydrochloride has long lasting antihypertensive action and reno-protective effect.

Lercanidipine hydrochloride is a lipophilic third-generation dihydropyridine-calcium channel blocker (DHP-CCB). Lercanidipine hydrochloride has long lasting antihypertensive action and reno-protective effect.

Specifications

Chemical Name or Material Lercanidipine hydrochloride
CAS 132866-11-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C36H42ClN3O6
Quantity 200 mg
Solubility Information DMSO : 50 mg/mL (77.14 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(C1=C(C)NC(C)=C(C(OC)=O)C1C2=CC=CC([N+]([O-])=O)=C2)OC(C)(C)CN(CCC(C3=CC=CC=C3)C4=CC=CC=C4)C.[H]Cl
Molecular Weight (g/mol) 648.19
Formula Weight 648.19
Percent Purity 99.94%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.