missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LEPREL1 Antibody [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20222JF585
This item is not returnable.
View return policy
Description
LEPREL1 Polyclonal antibody specifically detects LEPREL1 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| LEPREL1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Janelia Fluor 585 | |
| FLJ10718, leprecan-like 1, leprecan-like protein 1, MCVD, myxoid liposarcoma associated protein 4, myxoid liposarcoma-associated protein 4, P3H2, prolyl 3-hydroxylase 2, prolyl 3-hydroxylase 3 | |
| Produced in rabbits immunized with E. coli-derived Human LEPREL1 fragment | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
| 55214 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction