Learn More
Lepidine, 97%
Description
Lepidine is used in the preparation of certain dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lepidine is used in the preparation of certain dyes.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Incompatible with oxidizing agents. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 9°C to 10°C |
| Density | 1.083 |
| Boiling Point | 260°C to 263°C |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.62 |
| Quantity | 250 g |
| Beilstein | 110926 |
| Merck Index | 14,5441 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 143.19 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.