Learn More
LEO1 Mouse, anti-Human, Clone: 3D7A2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68568-1-IG-150UL
This item is not returnable.
View return policy
Description
The LEO1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LEO1. This antibody recognizes human antigen. The LEO1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| LEO1 | |
| 3D7A2 | |
| LEO1, RDL | |
| 150 μL | |
| 123169 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG1 |