Learn More
Emma Safety Footwear Leo XD Safety Shoes
Includes a steel toe cap, a polyurethane outer toe cap and an easy-twist sole that provides a sticky grip. Available in a variety of widths.
Brand: Emma Safety Footwear MM70156848
Additional Details : Weight : 1.91400kg
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather
- GripForce™ Easy Twist
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
Leo XD | |
Leather | |
48 |