Learn More
Invitrogen™ LentiArray™ CRISPR Negative Control Lentivirus, human, non-targeting
Description
Negative control gRNAs are essential in screening applications for setting thresholds for hit determination.
The Invitrogen™ LentiArray™ CRISPR Negative Control Lentivirus, Human Scrambled, contains a gRNA sequence with no sequence homology to any region of the human genome.
Recommended Storage
Store at -68 to -85°C
Specifications
Specifications
| Content And Storage | 1 x 100 μL. Store at -68 to -85°C. |
| Control Type | Non-targeting negative control |
| Delivery Type | Lentiviral |
| Promoter | U6 |
| Transfection Technique | Lentiviral transduction |
| Species | Human |
| For Use With (Application) | CRISPR gene knockout |
| Product Line | LentiArray |
| Product Type | CRISPR Negative Control Lentivirus |
| Quantity | 100 μL |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.