missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LentiArray™ Cas9 Lentivirus

Product Code. 15601477
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15601477 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. Not for use in diagnostic procedures.
Product Code. 15601477 Supplier Invitrogen™ Supplier No. A32069

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. Not for use in diagnostic procedures.

LentiArray Cas9 Lentivirus is provided as high-titer purified lentiviral particles and is an efficient method to drive high level Cas9 nuclease expression in a wide variety of cell types.

The LentiArray Cas9 Lentivirus construct includes a human codon-optimized version of Cas9 with two nuclear localization signals to facilitate efficient delivery into the nucleus. The 1-mL size is designed to support studies utilizing a co-infection approach (Cas9 lentivirus + guideRNA lentivirus delivered together) with either the LentiArray CRISPR libraries or LentiArray gRNAs against single gene targets. LentiArray Cas9 Lentivirus is provided as ready-to-use lentivirus at a titer of 1 x 107 TU/mL.

Features include:

  • 1 mL high-titer lentiviral particles
  • Ready-to-use lentiviral particles at a titer of 1 x 107 TU/mL
  • EF-1a promotor that is highly effective in a wide variety of cell models
  • Create stable Cas9-expressing cell models for screening and focused CRISPR knockout studies

Recommended Storage

Store at -68 to -85°C

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 x 1 mL. Store at -68 to -85°C.
Delivery Type Lentiviral
Sample Type siRNA
Promoter EF-1a
Transfection Technique Lentiviral transduction
Species Human
For Use With (Application) CRISPR gene knockout
Product Line LentiArray
Product Type Cas9 Lentivirus
Quantity 1 mL
Shipping Condition Dry Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.