missing translation for 'onlineSavingsMsg'
Learn More

Biochrom Lens for use with the Beamsplitter Assembly

Product Code. 15891232
Click to view available options
Type:
Plano Convex FL Lens
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15891232

Brand: Biochrom 80300701

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Plano Convex F/L Lens compatible with Biodrop Beam Splitter Assemblies.

  • Plano Convex F/L
  • 16 mm dia.
  • Compatible with Biodrop Beam Splitter (80-3005-41)

Specifications

Diameter (English) 0.6 in.
Diameter (Metric) 16 mm
Type Plano Convex FL Lens
Disposable Reusable
For Use With (Equipment) BioDrop Beam Splitter
Shape Plano Convex

For research use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.