Learn More
Stuart™ Lens Assembly
Click to view available options
For Use With (Equipment):
Stuart Digital Melting Apparatus (SMP10)
Unit Size:
Each
Description
- Samples tested in the SMP10 Digital Melting Apparatus are monitored through a detachable magnifying lens
- This replacement lens apparatus increases the cleanliness, and overall life of Stuart™ Melting Apparatuses
Specifications
Specifications
| Product Type | Replacement Lens Assembly |
| For Use With (Equipment) | Stuart Digital Melting Apparatus (SMP10) |