missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Lennox Luria-Bertani Broth

Product Code. 12881650
Click to view available options
Quantity:
500 g
2000 g
Unit Size:
2kg
500g
This item is not returnable. View return policy

Product Code. 12881650

Brand: Fisher BioReagents™ 12881650

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Used for the culturing of E. coli.

  • Large granules

Specifications

Media Type Lennox Luria-Bertani Broth
Product Type Microbiology Media
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.