missing translation for 'onlineSavingsMsg'
Learn More

Melford Lennox L Broth 20G Capsules

Product Code. 30018421
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
This item is not returnable. View return policy

Product Code. 30018421

Brand: Melford L240655000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

RPI media is now available in convenient premeasured and pre-mixed capsules.

RPI media is now available in convenient premeasured and pre-mixed capsules. Prebuffered media eliminates pH adjustments, dust and smells from the lab. Low Salt Formula. Molecular Biology Grade.

For propagation and maintenance of E.Coli:

Add 1 capsule per liter of water.

Heat and stir until capsule is completely dissolved.

Buffer is set for 7.2 at 20°C using a 10mM solution of TRIS/TRIS HCl

Sterilize by autoclaving for 15 minutes.

Ingredients:

Tryptone: 10 g/L

Yeast Extract: 5 g/L

Sodium Chloride: 5 g/L

Specifications

Quantity 5 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.