Learn More
Melford Lennox L Broth 20G Capsules
Description
RPI media is now available in convenient premeasured and pre-mixed capsules.
RPI media is now available in convenient premeasured and pre-mixed capsules. Prebuffered media eliminates pH adjustments, dust and smells from the lab. Low Salt Formula. Molecular Biology Grade.
For propagation and maintenance of E.Coli:
Add 1 capsule per liter of water.
Heat and stir until capsule is completely dissolved.
Buffer is set for 7.2 at 20°C using a 10mM solution of TRIS/TRIS HCl
Sterilize by autoclaving for 15 minutes.
Ingredients:
Tryptone: 10 g/L
Yeast Extract: 5 g/L
Sodium Chloride: 5 g/L
Specifications
Specifications
| Quantity | 1 kg |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.