missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lendalizumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA559284
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Lendalizumab, also known as an anti-C5 antibody, is a human monoclonal antibody targeting complement component 5 (C5). Complement component 5 (C5) is a vital protein in the complement system, part of the innate immune response. It is coded by the C5 gene located on chromosome 9, and consists of alpha and beta polypeptide chains. Activation of C5 occurs through the cleavage by C5 convertases in the classical, lectin, or alternative pathways, resulting in two fragments, C5a and C5b. C5a acts as an anaphylatoxin, promoting inflammation by stimulating chemotaxis and activating various immune cells. C5b initiates assembly of the membrane attack complex (MAC), leading to cell lysis. Dysregulation of C5 has been associated with autoimmune diseases such as myasthenia gravis and collagen-induced arthritis. Recent studies have explored the use of siRNA targeting C5 to mitigate disease symptoms by inhibiting C5 activation and subsequent inflammatory responses.
Specifications
| Lendalizumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG2SA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction