missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenampicillin hydrochloride, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
Unit Size:
10mg
2mg
5mg
Description
Lenampicillin hydrochloride (KBT 1585 hydrochloride) is an orally active prodrug of Ampicillin and is an effective beta-lactam antibacterial agent that inhibits bacterial penicillin-binding proteins (transpeptidase). Lenampicillin hydrochloride has improved absorption and decreased side effects compares to Ampicillin and is applied in the investigation of the suppurative skin and soft tissue infection .
Specifications
Specifications
| Chemical Name or Material | Lenampicillin hydrochloride |
| Physical Form | Solid |
| CAS | 80734-02-7 |
| Color | White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C21H24ClN3O7S |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | KBT 1585 hydrochloride |
| Solubility Information | DMSO : 100 mg/mL (200.82 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction