missing translation for 'onlineSavingsMsg'
Learn More

Lenampicillin hydrochloride, MedChemExpress

Product Code. 16410026
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
Unit Size:
10mg
2mg
5mg
This item is not returnable. View return policy

Product Code. 16410026

Brand: Medchem Express HY100500

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Lenampicillin hydrochloride (KBT 1585 hydrochloride) is an orally active prodrug of Ampicillin and is an effective beta-lactam antibacterial agent that inhibits bacterial penicillin-binding proteins (transpeptidase). Lenampicillin hydrochloride has improved absorption and decreased side effects compares to Ampicillin and is applied in the investigation of the suppurative skin and soft tissue infection .

Lenampicillin hydrochloride (KBT 1585 hydrochloride) is an orally active prodrug of Ampicillin and is an effective beta-lactam antibacterial agent that inhibits bacterial penicillin-binding proteins (transpeptidase). Lenampicillin hydrochloride has improved absorption and decreased side effects compares to Ampicillin and is applied in the investigation of the suppurative skin and soft tissue infection .

Specifications

Chemical Name or Material Lenampicillin hydrochloride
Physical Form Solid
CAS 80734-02-7
Color White
Quantity 2 mg
Purity Grade Notes Research
Molecular Formula C21H24ClN3O7S
For Use With (Application) Cancer-programmed cell death
Synonym KBT 1585 hydrochloride
Solubility Information DMSO : 100 mg/mL (200.82 mM; Need ultrasonic)
SMILES O=C([C@@H]1N(C2=O)[C@]([C@@H]2NC([C@@H](C3=CC=CC=C3)N)=O)([H])SC1(C)C)OCC(OC4=O)=C(O4)C.Cl
Molecular Weight (g/mol) 497.95
Formula Weight 497.95
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.