missing translation for 'onlineSavingsMsg'
Learn More

Lenalidomide hemihydrate, MedChemExpress

Product Code. 16461966
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16461966

Brand: Medchem Express HYA0003B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lenalidomide hemihydrate (CC-5013 hemihydrate), a derivative of Thalidomide, acts as molecular glue.

Lenalidomide hemihydrate is an orally active immunomodulator. Lenalidomide hemihydrate (CC-5013 hemihydrate) is a ligand of ubiquitin E3 ligase cereblon (CRBN), and it causes selective ubiquitination and degradation of two lymphoid transcription factors, IKZF1 and IKZF3, by the CRBN-CRL4 ubiquitin ligase. Lenalidomide hemihydrate (CC-5013 hemihydrate) specifically inhibits growth of mature B-cell lymphomas, including multiple myeloma, and induces IL-2 release from T cells.

Specifications

Chemical Name or Material Lenalidomide hemihydrate
CAS 847871-99-2
Color Pink
Purity Grade Notes Research
Molecular Formula C13H14N3O3.5
Quantity 500 mg
Synonym CC-5013 hemihydrate
Solubility Information DMSO : 50 mg/mL (186.38 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.37 mM; Need ultrasonic)
SMILES O=C1N(C2C(NC(CC2)=O)=O)CC3=C(N)C=CC=C31.[0.5 H2O]
Molecular Weight (g/mol) 268.27
Formula Weight 268.27
Percent Purity 98.8%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.