missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenalidomide hemihydrate, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Lenalidomide hemihydrate is an orally active immunomodulator. Lenalidomide hemihydrate (CC-5013 hemihydrate) is a ligand of ubiquitin E3 ligase cereblon (CRBN), and it causes selective ubiquitination and degradation of two lymphoid transcription factors, IKZF1 and IKZF3, by the CRBN-CRL4 ubiquitin ligase. Lenalidomide hemihydrate (CC-5013 hemihydrate) specifically inhibits growth of mature B-cell lymphomas, including multiple myeloma, and induces IL-2 release from T cells.
Specifications
Specifications
| Chemical Name or Material | Lenalidomide hemihydrate |
| CAS | 847871-99-2 |
| Color | Pink |
| Purity Grade Notes | Research |
| Molecular Formula | C13H14N3O3.5 |
| Quantity | 100 mg |
| Synonym | CC-5013 hemihydrate |
| Solubility Information | DMSO : 50 mg/mL (186.38 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.37 mM; Need ultrasonic) |
| SMILES | O=C1N(C2C(NC(CC2)=O)=O)CC3=C(N)C=CC=C31.[0.5 H2O] |
| Molecular Weight (g/mol) | 268.27 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction