missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ Lemo Polyplast Temp Electrode
Click to view available options
Length (Metric):
120 mm
Unit Size:
Each
Specifications
Specifications
| Accuracy (mV) | 0 ± 20mV |
| Accuracy (Temperature) | -10°C to +80°C |
| Diameter (Metric) | 12 mm |
| Electrical Requirements | 0 ± 20 mV, 57 to 59 mV |
| Length (Metric) | 120 mm |
| pH Range | 0 to 14 |
| Temperature (Metric) Range | -10°C to +80°C |
| Certifications/Compliance | Yes, with measured values in Buffer pH 4.01 and pH 7.00 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction