missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAN1 Monoclonal Antibody (OTI4E1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525021
This item is not returnable.
View return policy
Description
MAN1 Monoclonal Antibody for Western Blot, ICC/IF, IP
Can function as a specific repressor of TGF-beta, activin, and BMP signaling through its interaction with the R-SMAD proteins. Antagonizes TGF-beta-induced cell proliferation arrest.
Specifications
| MAN1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9Y2U8 | |
| LEMD3 | |
| Full length human recombinant protein of LEMD3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Western Blot, Immunocytochemistry, Western Blot | |
| OTI4E1 | |
| Unconjugated | |
| LEMD3 | |
| AI316861; inner nuclear membrane protein Man1; integral inner nuclear membrane protein; integral inner nuclear membrane protein MAN1; LEM domain containing 3; LEM domain-containing protein 3; LEMD3; MAN1; RGD1561001 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 23592 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction