missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LEMD3 Antibody (OTI6C10), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71744B
This item is not returnable.
View return policy
Description
LEMD3 Monoclonal specifically detects LEMD3 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| LEMD3 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| LEMD3 | |
| Full length recombinant protein of human LEMD3 (NP_055134) produced in HEK293T cells. | |
| 0.1 mL | |
| 23592.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI6C10 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| inner nuclear membrane protein Man1, integral inner nuclear membrane protein, LEM domain containing 3, MAN1LEM domain-containing protein 3 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction