missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LEMD3 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82104AF750
This item is not returnable.
View return policy
Description
LEMD3 Polyclonal antibody specifically detects LEMD3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| LEMD3 | |
| Polyclonal | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Rabbit | |
| Peptide Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 750 | |
| inner nuclear membrane protein Man1, integral inner nuclear membrane protein, LEM domain containing 3, MAN1LEM domain-containing protein 3 | |
| LEMD3 antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human LEMD3. The immunogen is located within the last 50 amino acids of LEMD3. Amino Acid Squence: CNTPLKPSNKHMNSMSHLR | |
| 0.1 mL | |
| Signal Transduction | |
| 23592 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction