missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LEMD2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82103
This item is not returnable.
View return policy
Description
LEMD2 Polyclonal specifically detects LEMD2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| LEMD2 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| LEMD2 | |
| LEMD2 antibody was raised against a 16 amino acid synthetic peptide near the center of human LEMD2. The immunogen is located within amino acids 220 - 270 of LEMD2. Amino Acid Squence: MGKPSAPQEAEDNMKL | |
| 0.1 mg | |
| Primary | |
| At least two isoforms of LEMD2 are known to exist; this antibody will detect only the longer isoform. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 2.5 μg/mL | |
| LEM domain containing 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 221496 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction