missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LEMD2 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-82103AF750
This item is not returnable.
View return policy
Description
LEMD2 Polyclonal antibody specifically detects LEMD2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| LEMD2 | |
| Polyclonal | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Rabbit | |
| Peptide Affinity-purified | |
| RUO | |
| 221496 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 750 | |
| LEM domain containing 2 | |
| LEMD2 antibody was raised against a 16 amino acid synthetic peptide near the center of human LEMD2. The immunogen is located within amino acids 220 - 270 of LEMD2. Amino Acid Squence: MGKPSAPQEAEDNMKL | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction