missing translation for 'onlineSavingsMsg'
Learn More

LEMD1 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18421907 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421907 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421907 Supplier Novus Biologicals Supplier No. NBP281920JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LEMD1 Polyclonal antibody specifically detects LEMD1 in Human, Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen LEMD1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias cancer/testis antigen 50, LEM domain containing 1, LEM domain-containing protein 1, LEMP-1
Host Species Rabbit
Immunogen LEMD1 antibody was raised against a 19 amino acid synthetic peptide near the center of human LEMD1. The immunogen is located within amino acids 90 - 140 of LEMD1. Amino Acid Squence: LDYKPSKGRRWAARAPST
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 93273
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.