missing translation for 'onlineSavingsMsg'
Learn More

Lemborexant, MedChemExpress

Product Code. 16489687
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489687 5 mg 5mg
16487158 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16489687 Supplier Medchem Express Supplier No. HY16725

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Lemborexant (E-2006) is a reversible, competitive and orally active dual antagonist of the orexin OX1 and OX2 receptors with IC50 values of 6.1 nM and 2.6 nM, respectively. Lemborexant can be treated insomnia.

Lemborexant (E-2006) is a reversible, competitive and orally active dual antagonist of the orexin OX1 and OX2 receptors with IC50 values of 6.1 nM and 2.6 nM, respectively. Lemborexant can be treated insomnia.

Specifications

Chemical Name or Material Lemborexant
CAS 1369764-02-2
Color White
Purity Grade Notes Research
Molecular Formula C22H20F2N4O2
Quantity 5 mg
Synonym E-2006
Solubility Information DMSO : 100 mg/mL (243.65 mM; Need ultrasonic)
SMILES O=C([C@H]1[C@@](C2=CC=CC(F)=C2)(COC3=CN=C(C)N=C3C)C1)NC4=NC=C(F)C=C4
Molecular Weight (g/mol) 410.42
Formula Weight 410.42
Percent Purity 99.19%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.