missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LEM3 Antibody, DyLight 488, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP2-41144G
This item is not returnable.
View return policy
Description
LEM3 Polyclonal antibody specifically detects LEM3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| LEM3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Chicken | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| DyLight 488 | |
| ANKRD41, ankyrin repeat and LEM domain containing 1, ankyrin repeat and LEM domain-containing protein 1, FLJ39369, LEM domain containing 6, LEMD6, LEM-domain containing protein 3 | |
| Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human ANKLE1. The immunogen is located within the last 50 amino acids of ANKLE1. Amino Acid Squence: CAGWPPARRRRLGVHL | |
| 0.1 mL | |
| Protein Kinase | |
| 126549 | |
| Store at 4°C in the dark. | |
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur