missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Leishman Stain

Product Code. 11464753
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464753 25 g 25g
11474753 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11464753 Supplier Thermo Scientific Chemicals Supplier No. A11644.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12627-53-1

Leishman Stain is used to stain blood smears to identify parasites, leukocytes and trypanosomas. It is used in hematology and tropical medicine to differentiate and identify leukocytes. It acts like fluorescent stains for detection of malaria parasite. It is also used for bone marrow staining.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Leishman Stain is used to stain blood smears to identify parasites, leukocytes and trypanosomas. It is used in hematology and tropical medicine to differentiate and identify leukocytes. It acts like fluorescent stains for detection of malaria parasite. It is also used for bone marrow staining.

Solubility
Soluble in water (4 mg/ml), methanol (1 mg/ml), ethanol (4 mg/ml), and methyl cellosolve (20 mg/ml).

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12627-53-1
MDL Number MFCD00131498

Specifications

Solubility Information Soluble in water (4mg/ml),methanol (1mg/ml),ethanol (4mg/ml),and methyl cellosolve (20mg/ml).
Odor Odorless
Quantity 25 g
Chemical Name or Material Leishman Stain

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.