missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Leishman Stain

Code produit 11474753
Change view
Click to view available options
Quantity:
25 g
100 g
Conditionnement:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity unitSize
11474753 100 g 100g
11464753 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11474753 Fournisseur Thermo Scientific Chemicals Code fournisseur A11644.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 12627-53-1

Leishman Stain is used to stain blood smears to identify parasites, leukocytes and trypanosomas. It is used in hematology and tropical medicine to differentiate and identify leukocytes. It acts like fluorescent stains for detection of malaria parasite. It is also used for bone marrow staining.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Leishman Stain is used to stain blood smears to identify parasites, leukocytes and trypanosomas. It is used in hematology and tropical medicine to differentiate and identify leukocytes. It acts like fluorescent stains for detection of malaria parasite. It is also used for bone marrow staining.

Solubility
Soluble in water (4 mg/ml), methanol (1 mg/ml), ethanol (4 mg/ml), and methyl cellosolve (20 mg/ml).

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 12627-53-1
MDL Number MFCD00131498

Spécification

Solubility Information Soluble in water (4mg/ml),methanol (1mg/ml),ethanol (4mg/ml),and methyl cellosolve (20mg/ml).
Odor Odorless
Quantity 100 g
Chemical Name or Material Leishman Stain
compliance-icons
Identifiant produit
  • Leishman Stain
Mention d'avertissement
  • Warning
Catégorie de danger
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Mention de danger
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .
  • P362-Take off contaminated clothing.
Informations supplémentaires
  • MIXTURE LIST-Contain: Leishman's stain

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.