missing translation for 'onlineSavingsMsg'
Learn More

Legumain/Asparaginyl Endopeptidase Rabbit anti-Human, Mouse, Clone: 6X5U6, Novus Biologicals™

Product Code. 18332992 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18332992 100 μg 100µL
18325212 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18332992 Supplier Novus Biologicals Supplier No. NBP316645100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Legumain/Asparaginyl Endopeptidase Monoclonal antibody specifically detects Legumain/Asparaginyl Endopeptidase in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Legumain/Asparaginyl Endopeptidase
Applications Western Blot
Classification Monoclonal
Clone 6X5U6
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias AEP, Asparaginyl endopeptidase, cysteine protease 1, legumain, LGMN1, Protease, cysteine 1, protease, cysteine, 1 (legumain), PRSC1EC 3.4.22.34
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 210-302 of human Legumain/Asparaginyl Endopeptidase (Q99538).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5641
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.