missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Legumain/Asparaginyl Endopeptidase Antibody (46I3) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£149.00 - £436.00
Specifications
| Antigen | Legumain/Asparaginyl Endopeptidase |
|---|---|
| Clone | 46I3 |
| Concentration | LYOPH |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18178414
|
Novus Biologicals
NBP2-21996-0.025MG |
0.025 mg |
£149.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18118641
|
Novus Biologicals
NBP2-21996 |
0.1 mg |
£436.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Legumain/Asparaginyl Endopeptidase Monoclonal specifically detects Legumain/Asparaginyl Endopeptidase in Human samples. It is validated for Western Blot, ELISA.Specifications
| Legumain/Asparaginyl Endopeptidase | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AEP, Asparaginyl endopeptidase, cysteine protease 1, legumain, LGMN1, Protease, cysteine 1, protease, cysteine, 1 (legumain), PRSC1EC 3.4.22.34 | |
| LGMN | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 46I3 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| Q99538 | |
| 5641 | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title