missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Legumain/Asparaginyl Endopeptidase Antibody (46I3), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-21996AF405
This item is not returnable.
View return policy
Description
Legumain/Asparaginyl Endopeptidase Monoclonal specifically detects Legumain/Asparaginyl Endopeptidase in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Legumain/Asparaginyl Endopeptidase | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LGMN | |
| Human recombinant Legumain/Asparaginyl Endopeptidase | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 46I3 | |
| Western Blot, ELISA | |
| AEP, Asparaginyl endopeptidase, cysteine protease 1, legumain, LGMN1, Protease, cysteine 1, protease, cysteine, 1 (legumain), PRSC1EC 3.4.22.34 | |
| Mouse | |
| Protein G purified | |
| Cancer | |
| 5641 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction