Learn More
Legionella pneumophila LPS Monoclonal Antibody (G90A), Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA518213
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This antibody is reactive with LPS of all L. pneumophila strains tested (11). Purity is >90%. Recommended pairs for ELISA: MA5-18213 (capture) and MA5-18214 (conjugated, detection).
The causative agent of what is commonly called Legionnaire's Disease, Legionella pneumophila causes acute pneumonia. The disease can also present itself as an nonpneumonic, influenza-like illness called Pontiac Fever. Legionella is found in aquatic environments where it's growth is supported by amoebae. In human infections Legionella acts as an intracellular parasite and multiplies in alveolar macrophages. The genus Legionella contains 39 recognized species. The immunological diversity of Legionella is due to the existence of a variety of serogroups. Legionella pneumophila has the most with a total of 14 serogroups.Specifications
Legionella pneumophila LPS | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG3 |
ELISA, Immunocytochemistry | |
G90A | |
Unconjugated | |
L. pneumophila; Legionellosis | |
Legionella pneumophila native protein | |
100 μg | |
Primary | |
4° C | |
Liquid |