missing translation for 'onlineSavingsMsg'
Learn More

Lefty-2 Antibody [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30737173 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30737173 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737173 Supplier Novus Biologicals Supplier No. AF7648MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Lefty-2 Polyclonal antibody specifically detects Lefty-2 in Mouse samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Spécification

Antigen Lefty-2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias EBAF, Endometrial Bleeding-Associated Factor, LEFTA, Left-Right Determination Factor 2, Left-right determination factor A, LEFTYA, Protein Lefty-2, Protein Lefty-A, TGFB4, TGF-beta-4, Transforming Growth Factor Beta-4
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Lefty-2, Phe78-Leu368, Accession # P57785
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Immunology, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 7044
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.