missing translation for 'onlineSavingsMsg'
Learn More

Lefty-2 Antibody [Biotin], Novus Biologicals™

Product Code. 30733837 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30733837 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733837 Supplier Novus Biologicals Supplier No. AF7648B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Lefty-2 Polyclonal antibody specifically detects Lefty-2 in Mouse samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lefty-2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias EBAF, Endometrial Bleeding-Associated Factor, LEFTA, Left-Right Determination Factor 2, Left-right determination factor A, LEFTYA, Protein Lefty-2, Protein Lefty-A, TGFB4, TGF-beta-4, Transforming Growth Factor Beta-4
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Lefty-2, Phe78-Leu368, Accession # P57785
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Immunology, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 7044
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.