missing translation for 'onlineSavingsMsg'
Learn More

Leflunomide, TRC

Product Code. 30478586
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478586 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478586 Supplier Toronto Research Chemicals Supplier No. TRCL32275050MG
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Leflunomide
CAS 75706-12-6
Molecular Formula C12 H9 F3 N2 O2
Quantity 50 mg
Synonym Leflunomide, 5-Methyl-N-[4-(trifluoromethyl)phenyl]-4-isoxazolecarboxamide, Arava, HWA 486, Leflunomida, Leflunomidum, SU 101
InChI Formula InChI=1S/C12H9F3N2O2/c1-7-10(6-16-19-7)11(18)17-9-4-2-8(3-5-9)12(13,14)15/h2-6H,1H3,(H,17,18)
SMILES Cc1oncc1C(=O)Nc2ccc(cc2)C(F)(F)F
IUPAC Name 5-methyl-N-[4-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxamide
Molecular Weight (g/mol) 270.21
Formula Weight 270.0616
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.