missing translation for 'onlineSavingsMsg'
Learn More

Leflunomide

Product Code. 15457129
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15457129 500 mg 500mg
15447129 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15457129 Supplier Thermo Scientific Alfa Aesar Supplier No. J65917.ME
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75706-12-6 | C12H9F3N2O2 | 270.211 g/mol

An immunosuppressive agent. Inhibits T and B cell proliferation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • An immunosuppressive agent
  • Inhibits T and B cell proliferation
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75706-12-6
Molecular Formula C12H9F3N2O2
Molecular Weight (g/mol) 270.211
MDL Number MFCD00867593
InChI Key VHOGYURTWQBHIL-UHFFFAOYSA-N
Synonym leflunomide, arava, leflunomidum, lefunamide, leflunomida, leflunomidum inn-latin, 5-methyl-n-4-trifluoromethyl phenyl-1,2-oxazole-4-carboxamide, leflunomida inn-spanish, su 101 pharmaceutical, arava tn
PubChem CID 3899
ChEBI CHEBI:6402
IUPAC Name 5-methyl-N-[4-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxamide
SMILES CC1=C(C=NO1)C(=O)NC2=CC=C(C=C2)C(F)(F)F

Specifications

Color White
Quantity 500 mg
UN Number UN2811
Merck Index 14,5432
Formula Weight 270.21
Physical Form Crystalline Solid
Chemical Name or Material Leflunomide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.