missing translation for 'onlineSavingsMsg'
Learn More

Leflunomide-d4, TRC

Product Code. 30466531
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30466531 10 mg 10mg
30471212 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30466531 Supplier Toronto Research Chemicals Supplier No. TRCL32275210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1189987-23-2
Molecular Formula C12 D4 H5 F3 N2 O2
Molecular Weight (g/mol) 274.232
Synonym 5-Methyl-N-[2,3,5,6-tetradeuterio-4-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxamide, 5-Methyl-N-[4-(trifluoromethyl)phenyl-2,3,5,6-d4]-4-isoxazolecarboxamide, Leflunomide-d4, Leflunomide-D4 (phenyl-D4)
IUPAC Name 5-methyl-N-[2,3,5,6-tetradeuterio-4-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxamide
SMILES [2H]c1c([2H])c(c([2H])c([2H])c1NC(=O)c2cnoc2C)C(F)(F)F

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C12H9F3N2O2/c1-7-10(6-16-19-7)11(18)17-9-4-2-8(3-5-9)12(13,14)15/h2-6H,1H3,(H,17,18)/i2D,3D,4D,5D
Formula Weight 274.087
Chemical Name or Material Leflunomide-d4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.