Learn More
LEF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14972-1-AP-150UL
 View more versions of this product
Product Code. 16873914
  • £349.00 / 150µL
Description
This gene encodes a transcription factor belonging to a family of proteins that share homology with the high mobility group protein-1. The protein encoded by this gene can bind to a functionally important site in the T-cell receptor-alpha enhancer, thereby conferring maximal enhancer activity. This transcription factor is involved in the Wnt signaling pathway, and it may function in hair cell differentiation and follicle morphogenesis. Mutations in this gene have been found in somatic sebaceous tumors. This gene has also been linked to other cancers, including androgen-independent prostate cancer. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
LEF1
Polyclonal
Unconjugated
LEF1
LEF 1, LEF1, TCF1 alpha, TCF1ALPHA
Rabbit
Antigen Affinity Chromatography
RUO
161452, 16842, 51176
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.41 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P27782, Q9QXN1, Q9UJU2
Lef1
LEF1 Fusion Protein Ag6882
150 μL
Primary
Human, Mouse, Rat