missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LEF1 Rabbit anti-Human, Clone: LEF1/341R, Novus Biologicals™
Rabbit Monoclonal Antibody
£235.00
Specifications
| Antigen | LEF1 |
|---|---|
| Clone | LEF1/341R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
Description
LEF1 Monoclonal specifically detects LEF1 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| LEF1 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| DNA Repair, DNA replication Transcription Translation and Splicing, Signal Transduction, Transcription Factors and Regulators, Wnt Signaling Pathway | |
| 10 mM PBS with 0.05% BSA | |
| 51176 | |
| IgG κ | |
| Protein A or G purified |
| LEF1/341R | |
| Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZp586H0919, FLJ46390, LEF-1, lymphoid enhancer-binding factor 1, T cell-specific transcription factor 1-alpha, TCF1-alpha, TCF1ALPHA | |
| Synthetic peptide corresponding to residues within aa100-200 of LEF1 | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title