missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LEF1 Monoclonal Antibody (E.281.8)
Rabbit Monoclonal Antibody
Brand: Invitrogen™ MA514966
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
This gene encodes a transcription factor belonging to a family of proteins that share homology with the high mobility group protein-1. The protein encoded by this gene can bind to a functionally important site in the T-cell receptor-alpha enhancer, thereby conferring maximal enhancer activity. This transcription factor is involved in the Wnt signaling pathway, and it may function in hair cell differentiation and follicle morphogenesis. Mutations in this gene have been found in somatic sebaceous tumors. This gene has also been linked to other cancers, including androgen-independent prostate cancer. Alternative splicing results in multiple transcript variants.
Specifications
| LEF1 | |
| Monoclonal | |
| 157 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 0.15M NaCl, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
| P27782, Q9QXN1, Q9UJU2 | |
| LEF1 | |
| Synthetic peptide corresponding to residues surrounding Pro82 of human LEF1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| E.281.8 | |
| Unconjugated | |
| LEF1 | |
| 3000002B05; AI451430; DKFZp586H0919; FLJ46390; HGNC:6551; HMG Binding Domain; LEF1; Lef-1; lymphoid enhancer binding factor 1; lymphoid enhancer binding factor 1 short isoform; lymphoid enhancer-binding factor 1; OTTHUMP00000219736; OTTHUMP00000219737; T cell-specific transcription factor 1-alpha; TCF10; TCF1ALPHA; TCF1-alpha; TCF7L3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 161452, 16842, 51176 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction