missing translation for 'onlineSavingsMsg'
Learn More

LEF1 Antibody (rLEF1/6906), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18347944 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18347944 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18347944 Supplier Novus Biologicals Supplier No. NBP314130AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LEF1 Monoclonal antibody specifically detects LEF1 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen LEF1
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rLEF1/6906
Conjugate Alexa Fluor 488
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DKFZp586H0919, FLJ46390, LEF-1, lymphoid enhancer-binding factor 1, T cell-specific transcription factor 1-alpha, TCF1-alpha, TCF1ALPHA
Host Species Mouse
Immunogen Recombinant fragment (around aa100-200) of human LEF1 protein (exact sequence is proprietary) (Uniprot: Q9UJU2)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing, Signal Transduction, Transcription Factors and Regulators, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 51176
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.