missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LEF1 Antibody (LEF1/6764), Novus Biologicals™
Mouse Monoclonal Antibody
£195.00 - £385.00
Specifications
| Antigen | LEF1 |
|---|---|
| Clone | LEF1/6764 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18621855
|
Novus Biologicals
NBP3-20278-20ug |
20 μg |
£195.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18690185
|
Novus Biologicals
NBP3-20278-100ug |
100 μg |
£385.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LEF1 Monoclonal antibody specifically detects LEF1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Spezifikation
| LEF1 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| DNA Repair, DNA replication Transcription Translation and Splicing, Signal Transduction, Transcription Factors and Regulators, Wnt Signaling Pathway | |
| 10mM PBS with 0.05% BSA | |
| 51176 | |
| IgG1 κ | |
| Protein A or G purified |
| LEF1/6764 | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZp586H0919, FLJ46390, LEF-1, lymphoid enhancer-binding factor 1, T cell-specific transcription factor 1-alpha, TCF1-alpha, TCF1ALPHA | |
| Recombinant fragment of human LEF1 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts