missing translation for 'onlineSavingsMsg'
Learn More

LEF1 Antibody (3H5), Novus Biologicals™

Product Code. 18336469 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18336469 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18336469 Supplier Novus Biologicals Supplier No. H00051176M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LEF1 Monoclonal antibody specifically detects LEF1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Proximity Ligation Assay, ELISA, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen LEF1
Applications Western Blot, ELISA, Immunocytochemistry, Proximity Ligation Assay, Sandwich ELISA, KnockDown
Classification Monoclonal
Clone 3H5
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH50632
Gene Alias DKFZp586H0919, FLJ46390, LEF-1, lymphoid enhancer-binding factor 1, T cell-specific transcription factor 1-alpha, TCF1-alpha, TCF1ALPHA
Host Species Mouse
Immunogen LEF1 (AAH50632, 1 a.a. ~ 399 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MPQLSGGGGGGGGDPELCATDEMIPFKDEGDPQKEKIFAEISHPEEEGDLADIKSSLVNESEIIPASNGHEVARQAQTSQEPYHDKAREHPDDGKHPDGGLYNKGPSYSSYSGYIMMPNMNNDPYMSNGSLSPPIPRTSNKVPVVQPSHAVHPLTPLITYSDEHFSPGSHPSHIPSDVNSKQGMSRHPPAPDIPTFYPLSPGGVGQITPPLGWQGQPVYPITGGFRQPYPSSLSVDTSMSRFSHHMIPGPPGPHTTGIPHPAIVTPQVKQEHPHTDSDLMHVKPQHEQRKEQEPKRPHIKKPLNAFMLYMKEMRANVVAECTLKESAAINQILGRRWHALSREEQAKYYELARKERQLHMQLYPGWSARDNYGKKKKRKREKLQESASGTGPRMTAAYI
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing, Signal Transduction, Transcription Factors and Regulators, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 51176
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.