missing translation for 'onlineSavingsMsg'
Learn More

Ledipasvir D-tartrate, MedChemExpress

Product Code. 16499227
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499227 5 mg 5mg
16466826 100 mg 100mg
16432757 50 mg 50mg
16416678 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499227 Supplier Medchem Express Supplier No. HY15602B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ledipasvir D-tartrate is an inhibitor of the hepatitis C virus NS5A, with EC50 values of 34 pM against GT1a and 4 pM against GT1b replicon.

Ledipasvir D-tartrate is an inhibitor of the hepatitis C virus NS5A, with EC50 values of 34 pM against GT1a and 4 pM against GT1b replicon.

Specifications

Chemical Name or Material Ledipasvir D-tartrate
CAS 1502654-87-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C53H60F2N8O12
Quantity 5 mg
Synonym GS-5885 D-tartrate
Solubility Information DMSO : ≥ 28 mg/mL (26.95 mM)
SMILES O=C(OC)N[C@H](C(N([C@H](C1=NC=C(C2=CC(C(F)(F)C3=C4C=CC(C5=CC=C6N=C([C@H]7N(C([C@@H](NC(OC)=O)C(C)C)=O)[C@]8([H])CC[C@@]7([H])C8)NC6=C5)=C3)=C4C=C2)N1)C9)CC%109CC%10)=O)C(C)C.O[C@@H]([C@@H](C(O)=O)O)C(O)=O
Molecular Weight (g/mol) 1039.09
Formula Weight 1039.09
Percent Purity 96.33%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.